All Photos(1)

109290

Sigma-Aldrich

Triethyl citrate

99%

Synonym(s):
Ethyl citrate
Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
Beilstein:
1801199
EC Number:
MDL number:
PubChem Substance ID:

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

assay

99%

refractive index

n20/D 1.442 (lit.)

bp

235 °C/150 mmHg (lit.)

solubility

peanut oil: soluble 0.8%
H2O: soluble ~6.9%
alcohol: miscible
diethyl ether: miscible

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethyl citrate is an eco-friendly plasticizer used in fabrication of green nanocomposites from cellulose acetate. It is added as plasticizer in the blend of poly(lactic acid) and starch in the presence of methylenediphenyl diisocyanate.

Application

Triethyl citrate was used for dye entrapment to prepare emission membrane filters for contact fluorescence imaging. It was used as plasticizer during coating of the HPMC caps and bodies with Aqoat AS-HF.

Certificate of Analysis

Certificate of Origin

Thermal and mechanical properties of poly (lactic acid)/starch/methylenediphenyl diisocyanate blending with triethyl citrate.
Ke T and Sun XS.
Journal of Applied Polymer Science, 88(13), 2947-2955 (2003)
Emilia Entcheva et al.
IEEE transactions on bio-medical engineering, 51(2), 333-341 (2004-02-10)
Tracking spatial and temporal determinants of cardiac arrhythmogenesis at the cellular level presents challenges to the optical mapping techniques employed. In this paper, we describe a compact system combining two nontraditional low-cost solutions for excitation light sources and emission filters...
Hwan-Man Park et al.
Biomacromolecules, 5(6), 2281-2288 (2004-11-09)
"Green" nanocomposites have been successfully fabricated from cellulose acetate (CA) powder, eco-friendly triethyl citrate (TEC) plasticizer and organically modified clay. The effect of the amount of plasticizer varying from 15 to 40 wt % on the performance of the nanocomposites...
Nathalie Huyghebaert et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 21(5), 617-623 (2004-04-07)
The aim of this study was to develop an alternative method for enteric coating of HPMC capsules that avoids the sealing step before coating, resulting in ready-to-use enteric-coated capsules for the use in retail or hospital pharmacy or R&D sections...
S Monfort et al.
International journal of food microbiology, 145(2-3), 476-482 (2011-02-19)
This paper is an evaluation of the lethal effectiveness of a successive application of pulsed electric fields (PEFs) and heat treatment in liquid whole egg (LWE) in the presence of different additives on the population of Salmonella serovar Enteritidis. Synergistic...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service