Documents

111899

Sigma-Aldrich

3-Methyl-3-ethylglutarimide

≥99%

Synonym(s):
3-Ethyl-3-methylglutarimide
Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
EC Number:
MDL number:
PubChem Substance ID:

assay

≥99%

mp

126-127 °C (lit.)

SMILES string

CCC1(C)CC(=O)NC(=O)C1

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service