Documents

127248

Sigma-Aldrich

4-Carbethoxy-2-ethyl-3-methyl-2-cyclohexen-1-one

technical grade

Synonym(s):
Ethyl 3-ethyl-2-methyl-4-oxo-2-cyclohexene-1-carboxylate, 2-Ethyl-Hagemann’s ester
Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27
EC Number:
MDL number:
PubChem Substance ID:

grade

technical grade

impurities

10-40% methyl ester

refractive index

n20/D 1.488 (lit.)

density

1.037 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C1CCC(=O)C(CC)=C1C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service