Skip to Content
MilliporeSigma

131997

Thymine

99%

Synonym(s):

2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
EC Number:
200-616-1
UNSPSC Code:
12352100
MDL number:
Beilstein/REAXYS Number:
117880
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

mp

~320 °C (dec.) (lit.)

SMILES string

CC1=CNC(=O)NC1=O

InChI

1S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)

InChI key

RWQNBRDOKXIBIV-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Thymine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library