134066

Sigma-Aldrich

o-Hydroxyhippuric acid

97%

Synonym(s):
2-Hydroxybenzoylaminoacetic acid, Salicylglycine
Empirical Formula (Hill Notation):
C9H9NO4
CAS Number:
Molecular Weight:
195.17
EC Number:
MDL number:

assay

97%

mp

167-169 °C (lit.)

SMILES string

OC(=O)CNC(=O)c1ccccc1O

InChI

1S/C9H9NO4/c11-7-4-2-1-3-6(7)9(14)10-5-8(12)13/h1-4,11H,5H2,(H,10,14)(H,12,13)

InChI key

ONJSZLXSECQROL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service