142980

Sigma-Aldrich

2-Chloro-5-(fluorosulfonyl)benzoic acid

Linear Formula:
ClC6H3(SO2F)CO2H
CAS Number:
Molecular Weight:
238.62
EC Number:
MDL number:

mp

148-150 °C (lit.)

SMILES string

OC(=O)c1cc(ccc1Cl)S(F)(=O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service