Documents

145483

Sigma-Aldrich

2,3-Diphenylquinoxaline

97%

Empirical Formula (Hill Notation):
C20H14N2
CAS Number:
Molecular Weight:
282.34
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

mp

125-127 °C (lit.)

SMILES string

c1ccc(cc1)-c2nc3ccccc3nc2-c4ccccc4

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.