MilliporeSigma
  • 150118
All Photos(1)

150118

Sigma-Aldrich

2,5-Dichlorophenylhydrazine

98%

Linear Formula:
Cl2C6H3NHNH2
CAS Number:
Molecular Weight:
177.03
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

98%

form

solid

mp

102-103 °C (lit.)

SMILES string

NNc1cc(Cl)ccc1Cl

InChI

1S/C6H6Cl2N2/c7-4-1-2-5(8)6(3-4)10-9/h1-3,10H,9H2

InChI key

LZKWWERBNXLGLI-UHFFFAOYSA-N

Application

2,5-Dichlorophenylhydrazine was used in the preparation of D-erythrose 2,5-dichlorophenylhydrazon.

Packaging

5, 25 g in glass bottle

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service