150142

Sigma-Aldrich

Fluorene-2-carboxaldehyde

99%

Empirical Formula (Hill Notation):
C14H10O
CAS Number:
Molecular Weight:
194.23
Beilstein/REAXYS Number:
2255852
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

assay

99%

mp

83-85 °C (lit.)

SMILES string

[H]C(=O)c1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C14H10O/c15-9-10-5-6-14-12(7-10)8-11-3-1-2-4-13(11)14/h1-7,9H,8H2

InChI key

MNQGEQSXFDKAPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorene-2-carboxaldehyde was used in the preparation of 2-hydroxyl-methyl-fluorene.

Packaging

5 g in glass bottle

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Sabeel M Basheer et al.
Materials science & engineering. C, Materials for biological applications, 72, 667-675 (2016-12-28)
1,5-Bis(2-fluorene)thiocarbohydrazone (FBTC) was designed and synthesized for selective sensing of fluoride and copper ions. The binding constants of FBTC towards fluoride and copper ions have been calculated using the Benesi-Hildebrand equation, and FBTC has more binding affinity towards copper ion...
Soundrapandian Suganthi et al.
Journal of biomolecular structure & dynamics, 37(13), 3305-3313 (2018-08-18)
G-quadruplex-binders have the plausible potential to act as anticancer agents. Herein, the mode of binding of a synthesized fluorenyl derivative of β-cyclodextrin with a duplex and G-quadrulex DNAs has been investigated. Moreover, the loading of the well-known G-quadruplex binder, berberine...
A J Dobson et al.
Acta crystallographica. Section C, Crystal structure communications, 54 ( Pt 12), 1890-1892 (1999-01-28)
The title compound, C14H10O, crystallized in the centrosymmetric space group P2(1)/c with a single molecule in the asymmetric unit. The C-H...O interaction having the shortest C...O distance involves the ring-bridging C9 atom and one of its H atoms, and results...
Synthesis and photophysical properties of porphyrins with fluorenyl pendant arms.
Drouet S, et al.
Tetrahedron, 65(15), 2975-2981 (2009)
Joëlle Akl et al.
Dalton transactions (Cambridge, England : 2003), 43(33), 12721-12733 (2014-07-12)
The 4'-(2-fluorenyl)-2,2':6',2''-terpyridine (FT) ligand and its cis(Cl,Cl)- and trans(Cl,Cl)-[Ru(II)(FT)Cl2(NO)](PF6) complexes have been synthesized. Both isomers were separated by HPLC and fully characterized by (1)H and (13)C NMR. The X-ray diffraction crystal structures were solved for FT (Pna21 space group, a...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.