15454

Sigma-Aldrich

Boc-Leu-OSu

Synonym(s):
N-(tert-Butoxycarbonyl)-L-leucine N-hydroxysuccinimide ester, Boc-L-leucine hydroxysuccinimide ester
Empirical Formula (Hill Notation):
C15H24N2O6
CAS Number:
Molecular Weight:
328.36
Beilstein/REAXYS Number:
4538026
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

≥99.0% (TLC)

optical activity

[α]20/D −43.5±1°, c = 1% in dioxane

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis: suitable

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

InChI

1S/C15H24N2O6/c1-9(2)8-10(16-14(21)22-15(3,4)5)13(20)23-17-11(18)6-7-12(17)19/h9-10H,6-8H2,1-5H3,(H,16,21)/t10-/m0/s1

InChI key

WXRGJQZMGGGTSS-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

5 g in glass bottle

Other Notes

Used for the quantative determination of D- and L-amino acids

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
A. Mitchell et al.
Analytical Chemistry, 50, 637-637 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.