15476

Sigma-Aldrich

Boc-D-Cha-OH hydrate

~1 mol/mol water

Synonym(s):
Boc-3-cyclohexyl-D-alanine hydrate, Boc-hexahydro-D-phenylalanine
Empirical Formula (Hill Notation):
C14H25NO4 · xH2O
CAS Number:
Molecular Weight:
271.35 (anhydrous basis)
Beilstein/REAXYS Number:
4192671
MDL number:
PubChem Substance ID:

optical activity

[α]20/D +15.5±1°, c = 2% in glacial acetic acid

impurities

~1 mol/mol water

mp

64-67 °C

SMILES string

O.CC(C)(C)OC(=O)N[C@H](CC1CCCCC1)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service