Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

155063

Sigma-Aldrich

1-Decalone,mixture of cis and trans

97%

Synonym(s):

Decahydro-1-naphthalenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

73-75 °C/1 mmHg (lit.)

density

0.986 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

O=C1CCCC2CCCCC12

InChI

1S/C10H16O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h8-9H,1-7H2

InChI key

AFEFRXAPJRCTOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-1-decalone was used as a substrate for the ketoreductase of the diketide synthase[1]. cis-1-Decalone was used in the preparation of [cis-cis-1-decalyl glucosid]-uronic acid[2].

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

190.4 °F - closed cup

flash_point_c

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service