15516

Sigma-Aldrich

Boc-Trp-OSu

Synonym(s):
Nα-Boc-L-tryptophan hydroxysuccinimide ester
Empirical Formula (Hill Notation):
C20H23N3O6
CAS Number:
Molecular Weight:
401.41
Beilstein/REAXYS Number:
462939
EC Number:
MDL number:
PubChem Substance ID:

optical activity

[α]20/D −24.0±2.0°, c = 1% in dioxane

application(s)

peptide synthesis: suitable

mp

146-148 °C

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)ON3C(=O)CCC3=O

InChI

1S/C20H23N3O6/c1-20(2,3)28-19(27)22-15(18(26)29-23-16(24)8-9-17(23)25)10-12-11-21-14-7-5-4-6-13(12)14/h4-7,11,15,21H,8-10H2,1-3H3,(H,22,27)/t15-/m0/s1

InChI key

CPJXMXQYRHNIFU-HNNXBMFYSA-N

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.