MilliporeSigma
All Photos(1)

15559

Sigma-Aldrich

Boc-His(Tos)-OH

≥98.0% (TLC)

Synonym(s):
Nα-Boc-N(im)-tosyl-L-histidine
Empirical Formula (Hill Notation):
C18H23N3O6S
CAS Number:
Molecular Weight:
409.46
Beilstein:
769957
MDL number:
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (TLC)

optical activity

[α]20/D +16±1°, c = 1% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

~125 °C (dec.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)n2cnc(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)c2

InChI

1S/C18H23N3O6S/c1-12-5-7-14(8-6-12)28(25,26)21-10-13(19-11-21)9-15(16(22)23)20-17(24)27-18(2,3)4/h5-8,10-11,15H,9H2,1-4H3,(H,20,24)(H,22,23)/t15-/m0/s1

InChI key

DCLJSEPKYJSEHW-HNNXBMFYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service