Skip to Content
MilliporeSigma

159964

1-Phenyl-1-cyclopropanecarbonitrile

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5C3H4CN
CAS Number:
Molecular Weight:
143.19
UNSPSC Code:
12352100
EC Number:
213-304-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

refractive index

n20/D 1.539 (lit.)

bp

133-137 °C/30 mmHg (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

N#CC1(CC1)c2ccccc2





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library