MilliporeSigma
All Photos(1)

160555

Sigma-Aldrich

1-Cyanoisourea sodium salt

98%

Linear Formula:
H2NC(ONa)=NCN
CAS Number:
Molecular Weight:
107.05
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

>300 °C (lit.)

SMILES string

[Na+].N\C([O-])=N\C#N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service