All Photos(1)

162159

Sigma-Aldrich

3-Cyclohexene-1,1-dimethanol

98%

Synonym(s):
4,4-Bis(hydroxymethyl)-1-cyclohexene, 3-Cyclohexene-1-ylidenedimethanol
Linear Formula:
C6H8(CH2OH)2
CAS Number:
Molecular Weight:
142.20
Beilstein:
2040659
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

88-90 °C (lit.)

SMILES string

OCC1(CO)CCC=CC1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service