163740

Sigma-Aldrich

EPPS

99%

Synonym(s):
N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulfonic acid), HEPPS, 4-(2-Hydroxyethyl)piperazine-1-propanesulfonic acid, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid
Empirical Formula (Hill Notation):
C9H20N2O4S
CAS Number:
Molecular Weight:
252.33
Beilstein/REAXYS Number:
3957385
EC Number:
MDL number:

assay

99%

useful pH range

7.3 - 8.7

pKa 

8

mp

237-239 °C (lit.)

SMILES string

OCCN1CCN(CCCS(O)(=O)=O)CC1

InChI

1S/C9H20N2O4S/c12-8-7-11-5-3-10(4-6-11)2-1-9-16(13,14)15/h12H,1-9H2,(H,13,14,15)

InChI key

OWXMKDGYPWMGEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Buffer useful in the physiological pH range.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service