163910

Sigma-Aldrich

3-Carbamoyl-PROXYL

97%

Synonym(s):
3-Carbamoyl-2,2,5,5-tetramethyl-1-pyrrolidineoxy, 3-Carbamoyl-2,2,5,5-tetramethylpyrrolidin-1-yloxy, free radical
Empirical Formula (Hill Notation):
C9H17N2O2
CAS Number:
Molecular Weight:
185.24
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

mp

173-175 °C (lit.)

SMILES string

CC1(C)CC(C(N)=O)C(C)(C)N1[O]

InChI

1S/C9H17N2O2/c1-8(2)5-6(7(10)12)9(3,4)11(8)13/h6H,5H2,1-4H3,(H2,10,12)

InChI key

XNNPAWRINYCIHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A spin label useful for non-invasive detection of free radicals in vivo by electron spin resonance imaging.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service