17013

Sigma-Aldrich

N,N″-Di-Z-diethylenetriamine

≥97.0% (HPLC)

Synonym(s):
Dibenzyl 2,2′-iminobis(ethylcarbamate)
Empirical Formula (Hill Notation):
C20H25N3O4
CAS Number:
Molecular Weight:
371.43
Beilstein/REAXYS Number:
7083538
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

assay

≥97.0% (HPLC)

reaction suitability

reagent type: cross-linking reagent

mp

70-74 °C

SMILES string

O=C(NCCNCCNC(=O)OCc1ccccc1)OCc2ccccc2

InChI

1S/C20H25N3O4/c24-19(26-15-17-7-3-1-4-8-17)22-13-11-21-12-14-23-20(25)27-16-18-9-5-2-6-10-18/h1-10,21H,11-16H2,(H,22,24)(H,23,25)

InChI key

DWPBEWIGNADCAX-UHFFFAOYSA-N

Packaging

5 g in poly bottle
1 g in glass bottle

Other Notes

Building block to introduce e.g. a protected 2,2′-iminodi(ethylamine) scaffold in libraries

Signal Word

Danger

Hazard Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3077 9 / PGIII

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
M. Adamczyk et al.
Organic preparations and procedures international, 30, 339-339 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.