17250

Sigma-Aldrich

2-{1-4-(Hydroxymethyl)phenylamino-3-methylbutylidene}-5,5-dimethyl-1,3-cyclohexanedione

≥98.0% (HPLC)

Synonym(s):
Dmab-OH
Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
MDL number:
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

assay

≥98.0% (HPLC)

mp

156-160

storage temp.

2-8°C

SMILES string

CC(C)C\C(Nc1ccc(CO)cc1)=C2/C(=O)CC(C)(C)CC2=O

InChI

1S/C20H27NO3/c1-13(2)9-16(21-15-7-5-14(12-22)6-8-15)19-17(23)10-20(3,4)11-18(19)24/h5-8,13,21-22H,9-12H2,1-4H3

InChI key

GARPDGDSXTZYQQ-UHFFFAOYSA-N

Related Categories

Application

2-{1-4-(Hydroxymethyl)phenylamino-3-methylbutylidene}-5,5-dimethyl-1,3-cyclohexanedione was used to protect glutamic and aspartic acids.

Packaging

2.5 g in poly bottle

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Bifunctional ligands based on the DOTA-monoamide cage.
Barge A, et al.
Organic & Biomolecular Chemistry, 1176-1184 (2008)
Aurélie Morin et al.
Cell reports, 30(13), 4551-4566 (2020-04-03)
Loss-of-function mutations in the SDHB subunit of succinate dehydrogenase predispose patients to aggressive tumors characterized by pseudohypoxic and hypermethylator phenotypes. The mechanisms leading to DNA hypermethylation and its contribution to SDH-deficient cancers remain undemonstrated. We examine the genome-wide distribution of 5-methylcytosine and...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.