187224

Sigma-Aldrich

2-Naphthalenesulfonic acid, magnesium salt-bis(hexachlorocyclopentadiene) adduct

technical grade

Empirical Formula (Hill Notation):
C40H14Cl24MgO6S2
CAS Number:
Molecular Weight:
1529.84
MDL number:

grade

technical grade

SMILES string

[Mg++].[O-]S(=O)(=O)c1ccc2C3C(C4C(c2c1)[C@@]5(Cl)C(Cl)=C(Cl)[C@]4(Cl)C5(Cl)Cl)[C@@]6(Cl)C(Cl)=C(Cl)[C@]3(Cl)C6(Cl)Cl.[O-]S(=O)(=O)c7ccc8C9C(C%10C(c8c7)[C@]%11(Cl)C(Cl)=C(Cl)[C@@]%10(Cl)C%11(Cl)Cl)[C@]%12(Cl)C(Cl)=C(Cl)[C@@]9(Cl)C%12(Cl)Cl

Application

Precursor to substituted naphthalenes which are otherwise difficult to synthesize.

Packaging

Packaged in poly bottles

Certificate of Analysis

Certificate of Origin

Aldrichimica Acta, 7, 23-23 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service