190462

Sigma-Aldrich

N-Methallyl-N′-methylthiourea

98%

Synonym(s):
N-Methyl-N′-(2-methyl-2-propen-1-yl)thiourea
Linear Formula:
H2C=C(CH3)CH2NHCSNHCH3
CAS Number:
Molecular Weight:
144.24
Beilstein/REAXYS Number:
2349725
EC Number:
MDL number:

assay

98%

mp

60-62 °C

SMILES string

CNC(=S)NCC(C)=C

Looking for similar products? Visit Product Comparison Guide

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service