190659

Sigma-Aldrich

α-Acetylphenylacetonitrile

98%

Synonym(s):
2-Phenylacetoacetonitrile
Linear Formula:
C6H5CH(COCH3)CN
CAS Number:
Molecular Weight:
159.18
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

92-94 °C (lit.)

SMILES string

CC(=O)C(C#N)c1ccccc1

Pictograms

Exclamation mark

Signal Word

Warning

hazcat

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

storage_class_code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service