Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19409

Sigma-Aldrich

1,3-Dimethylimidazolium methyl sulfate

≥98.5% (HPLC)

Synonym(s):

DMIMMeSO4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S
CAS Number:
Molecular Weight:
208.24
Beilstein/REAXYS Number:
9352735
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98.5% (HPLC)

form

solid

impurities

≤500 ppm water

density

1.33 g/mL at 20 °C (lit.)

SMILES string

COS([O-])(=O)=O.Cn1cc[n+](C)c1

InChI

1S/C5H9N2.CH4O4S/c1-6-3-4-7(2)5-6;1-5-6(2,3)4/h3-5H,1-2H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

WOKQGMYCUGJNIJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service