Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

195189

Sigma-Aldrich

Mordant Brown 33

Dye content 60 %

Synonym(s):

2,4-Diamino-5-(2-hydroxy-5-nitrophenylazo)benzenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H2[N=NC6H3(OH)NO2]SO3Na
CAS Number:
Molecular Weight:
375.29
Colour Index Number:
13250
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 60%

λmax

442 nm

SMILES string

[Na+].Nc1cc(N)c(cc1\N=N\c2cc(ccc2O)[N+]([O-])=O)S([O-])(=O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service