Documents

195448

Sigma-Aldrich

6-Methoxytryptamine

99%

Synonym(s):
3-(2-Aminoethyl)-6-methoxyindole
Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
EC Number:
MDL number:

assay

99%

mp

146-147 °C (lit.)

SMILES string

COc1ccc2c(CCN)c[nH]c2c1

InChI

1S/C11H14N2O/c1-14-9-2-3-10-8(4-5-12)7-13-11(10)6-9/h2-3,6-7,13H,4-5,12H2,1H3

InChI key

VOCGEKMEZOPDFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service