196053

Sigma-Aldrich

(−)-Carvyl acetate

≥98%

Synonym(s):
(1RS,5R)-5-Isopropenyl-2-methyl-2-cyclohexenyl acetate
Empirical Formula (Hill Notation):
C12H18O2
CAS Number:
Molecular Weight:
194.27
Beilstein/REAXYS Number:
2330666
EC Number:
MDL number:

assay

≥98%

optical activity

[α]23/D −81.6°, neat

refractive index

n20/D 1.475 (lit.)

bp

77-79 °C/0.1 mmHg (lit.)

density

0.972 g/mL at 20 °C (lit.)
0.976 g/mL at 25 °C

SMILES string

CC(=C)C1CC=C(C)C(C1)OC(C)=O

InChI

1S/C12H18O2/c1-8(2)11-6-5-9(3)12(7-11)14-10(4)13/h5,11-12H,1,6-7H2,2-4H3

InChI key

YTHRBOFHFYZBRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service