197505

Sigma-Aldrich

2,3-Cyclododecenopyridine

≥98%

Synonym(s):
5,6,7,8,9,10,11,12,13,14-Decahydrocyclododeca[b]pyridine
Empirical Formula (Hill Notation):
C15H23N
CAS Number:
Molecular Weight:
217.35
EC Number:
MDL number:

assay

≥98%

refractive index

n20/D 1.538 (lit.)

bp

165-175 °C/3.7 mmHg (lit.)

mp

22-23 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

C1CCCCCc2ncccc2CCCC1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

hazcat

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

storage_class_code

10 - Combustible liquids

WGK Germany

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service