MilliporeSigma
All Photos(1)

201197

Sigma-Aldrich

Beryllium chloride

purified by sublimation, 99%

Empirical Formula (Hill Notation):
BeCl2
CAS Number:
Molecular Weight:
79.92
EC Number:
MDL number:
PubChem Substance ID:

assay

99%

purified by

sublimation

mp

399 °C (lit.)

density

1.899 g/mL at 25 °C (lit.)

SMILES string

Cl[Be]Cl

InChI

1S/Be.2ClH/h;2*1H/q+2;;/p-2

InChI key

LWBPNIJBHRISSS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for novel sterically encumbered arylberyllium compounds.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service