All Photos(1)

202819

Sigma-Aldrich

Bismuth

powder, −100 mesh, ≥99.999%

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
EC Number:
MDL number:
PubChem Substance ID:

vapor pressure

<0.1 mmHg ( 20 °C)

assay

≥99.999%

form

powder

resistivity

129 μΩ-cm, 20°C

particle size

−100 mesh

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Elissa S Finkler et al.
Foot & ankle international, 36(11), 1310-1315 (2015-08-01)
The traditional nonsurgical accommodative treatment for diabetes-associated Charcot foot arthropathy has been unsuccessful in improving the quality of life in affected individuals. This has led to the growing interest in surgical correction of the acquired deformity with either "super construct"...
Andreia M C Rocha et al.
British journal of haematology, 168(3), 421-428 (2014-09-27)
Helicobacter pylori eradication induces platelet recovery in a subgroup of patients with chronic immune thrombocytopenia (cITP), but the mechanisms involved are still not understood. We aimed to evaluate the effect of H. pylori eradication on platelet response and to identify the...
Yifan Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(11), 3211-3216 (2015-03-05)
Glutathione and multidrug resistance protein (MRP) play an important role on the metabolism of a variety of drugs. Bismuth drugs have been used to treat gastrointestinal disorder and Helicobacter pylori infection for decades without exerting acute toxicity. They were found...
Emma Aneheim et al.
Scientific reports, 5, 12025-12025 (2015-07-15)
To facilitate multicentre clinical studies on targeted alpha therapy, it is necessary to develop an automated, on-site procedure for conjugating rare, short-lived, alpha-emitting radionuclides to biomolecules. Astatine-211 is one of the few alpha-emitting nuclides with appropriate chemical and physical properties...
Ivan A Popov et al.
Nature communications, 6, 8654-8654 (2015-10-13)
The electron deficiency and strong bonding capacity of boron have led to a vast variety of molecular structures in chemistry and materials science. Here we report the observation of highly symmetric cobalt-centered boron drum-like structures of CoB16(-), characterized by photoelectron...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service