Documents

206024

Sigma-Aldrich

Platinum on carbon

extent of labeling: 0.5 wt. % loading, 4-8 mesh

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
PubChem Substance ID:

extent of labeling

0.5 wt. % loading

particle size

4-8 mesh

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service