206334

Sigma-Aldrich

Evans Blue

Dye content 85 %

Synonym(s):
Direct Blue 53
Empirical Formula (Hill Notation):
C34H24N6Na4O14S4
CAS Number:
Molecular Weight:
960.81
Colour Index Number:
23860
EC Number:
MDL number:
PubChem Substance ID:

composition

Dye content, 85%

λmax

611 nm

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1\N=N\c2ccc3c(cc(c(N)c3c2O)S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(\N=N\c5ccc6c(cc(c(N)c6c5O)S([O-])(=O)=O)S([O-])(=O)=O)c(C)c4

InChI

1S/C34H28N6O14S4.4Na/c1-15-11-17(3-7-21(15)37-39-23-9-5-19-25(55(43,44)45)13-27(57(49,50)51)31(35)29(19)33(23)41)18-4-8-22(16(2)12-18)38-40-24-10-6-20-26(56(46,47)48)14-28(58(52,53)54)32(36)30(20)34(24)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4/b39-37+,40-38+;;;;

InChI key

ATNOAWAQFYGAOY-GPTZEZBUSA-J

Gene Information

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Cynthia N Perry et al.
Methods in enzymology, 453, 325-342 (2009-02-17)
Autophagy, a highly conserved cellular mechanism wherein various cellular components are broken down and recycled through lysosomes, occurs constitutively in the heart and may serve as a cardioprotective mechanism in some situations. It has been implicated in the development of...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service