MilliporeSigma
All Photos(1)

208116

Sigma-Aldrich

Sodium oxide

97%

Linear Formula:
Na2O
CAS Number:
Molecular Weight:
61.98
EC Number:
MDL number:

assay

97%

density

2.27 g/mL at 25 °C (lit.)

SMILES string

[O--].[Na+].[Na+]

InChI

1S/2Na.O/q2*+1;-2

InChI key

KKCBUQHMOMHUOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service