MilliporeSigma
All Photos(1)

211478

Sigma-Aldrich

1-Bromo-2,4-dimethylbenzene

97%

Synonym(s):
4-Bromo-m-xylene
Linear Formula:
(CH3)2C6H3Br
CAS Number:
Molecular Weight:
185.06
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

97%

form

liquid

refractive index

n20/D 1.551 (lit.)

bp

214 °C (lit.)

density

1.37 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Br)c(C)c1

InChI

1S/C8H9Br/c1-6-3-4-8(9)7(2)5-6/h3-5H,1-2H3

InChI key

YSFLQVNTBBUKEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Gold-catalyzed coupling reaction of pyrazine with 1-bromo-2,4-dimethylbenzene has been reported. 1-Bromo-2,4-dimethylbenzene reacts with Mg and DMF, followed by the reaction with I2 and aq.NH3 to afford the corresponding aromatic nitrile.

Packaging

25 g in glass bottle

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service