216895

Aminophyllin hydrate

99%

Synonym(s):
Theophylline, compound with ethylenediamine (2:1) dihydrate, 3,7Dihydro1,3dimethyl1Hpurine2,6dione, Cmpd. with 1,2ethanediamine (2:1), (Theophylline)2 ªO Ethylenediamine, Theophyllin hemiethylenediamine complex
Empirical Formula (Hill Notation):
C14H16N8O4 · C2H8N2 · xH2O
CAS Number:
Molecular Weight:
420.43 (anhydrous basis)
EC Number:
MDL number:

assay

99%

SMILES string

NCCN.CN1C(=O)N(C)c2nc[nH]c2C1=O.CN3C(=O)N(C)c4nc[nH]c4C3=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service