220086

Sigma-Aldrich

DL-Isocitric acid trisodium salt hydrate

97%

Synonym(s):
threo-DsLs-Isocitric acid trisodium salt hydrate
Empirical Formula (Hill Notation):
C6H5Na3O7 · xH2O
CAS Number:
Molecular Weight:
258.07 (anhydrous basis)
MDL number:
PubChem Substance ID:

assay

97%

SMILES string

O.[Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.H2O/c7-3(8)1-2(5(10)11)4(9)6(12)13;;;;/h2,4,9H,1H2,(H,7,8)(H,10,11)(H,12,13);;;;1H2/q;3*+1;/p-3

InChI key

BPEHKAXIRZOKIS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

An-Ping Lin et al.
Biochemistry, 50(38), 8241-8250 (2011-08-25)
Yeast NAD(+)-specific isocitrate dehydrogenase is an allosterically regulated octameric enzyme composed of four heterodimers of a catalytic IDH2 subunit and a regulatory IDH1 subunit. Despite structural predictions that the enzyme would contain eight isocitrate binding sites, four NAD(+) binding sites...
Chul-Sa Kim et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 63(9-10), 755-760 (2008-12-02)
Eight compounds isolated from Indian barnyard millet have been identified as L-malic acid, trans-aconitic acid, (+)-isocitric acid, 5-O-caffeoylquinic acid, 4-O-caffeoylquinic acid, isocarlinoside, 2"-O-rhamnosylisoorientin, and 7-O-(2"-O-glucuronosyl)glucuronosyltricin, respectively. These compounds showed high antifeeding activity against brown planthopper only when they were combined.
Mutations to metabolic enzymes in cancer herald a need to unify genetics and biochemistry.
Akane Kawamura et al.
Cell cycle (Georgetown, Tex.), 10(17), 2819-2820 (2011-08-23)
Grant C Bullock et al.
Blood, 116(1), 97-108 (2010-04-22)
Human red cell differentiation requires the action of erythropoietin on committed progenitor cells. In iron deficiency, committed erythroid progenitors lose responsiveness to erythropoietin, resulting in hypoplastic anemia. To address the basis for iron regulation of erythropoiesis, we established primary hematopoietic...
Ming-dong Wang et al.
Chinese medical journal, 124(17), 2611-2615 (2011-11-02)
Site A132Arg mutations potentially impair the affinity of isocitrate dehydrogenase 1 (IDH1) for its substrate isocitrate (ICT), consequently reducing the production of α-ketoglutarate and leading to tumor growth through the induction of the hypoxia-inducible factor-1 (HIF-1) pathway. However, given that...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service