221317

Sigma-Aldrich

Potassium hydrogenphosphate trihydrate

≥99%

Synonym(s):
Potassium phosphate dibasic trihydrate, sec.-Potassium phosphate, Dipotassium hydrogen phosphate trihydrate
Linear Formula:
K2HPO4 · 3H2O
CAS Number:
Molecular Weight:
228.22
EC Number:
MDL number:

assay

≥99%

SMILES string

O.O.O.[K+].[K+].OP([O-])([O-])=O

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service