Documents

223174

Sigma-Aldrich

Ethyl stearate

99%

Synonym(s):
Stearic acid ethyl ester, Ethyl octadecanoate
Linear Formula:
CH3(CH2)16COOC2H5
CAS Number:
Molecular Weight:
312.53
Beilstein/REAXYS Number:
1788183
EC Number:
MDL number:

assay

99%

bp

213-215 °C/15 mmHg (lit.)

mp

34-38 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC

InChI

1S/C20H40O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h3-19H2,1-2H3

InChI key

MVLVMROFTAUDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

WGK Germany

2

Flash Point(F)

235 °F

Flash Point(C)

113 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.