Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

225010

Flavianic acid hydrate

97%

Synonym(s):

8-Hydroxy-5,7-dinitro-2-naphthalenesulfonic acid

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
HOC10H4(NO2)2SO3H · xH2O
CAS Number:
Molecular Weight:
314.23 (anhydrous basis)
UNSPSC Code:
12171500
PubChem Substance ID:
EC Number:
207-600-3
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

O.Oc1c(cc([N+]([O-])=O)c2ccc(cc12)S(O)(=O)=O)[N+]([O-])=O

assay

97%

mp

148-151 °C (lit.)

λmax

390 nm (2nd), 428 nm

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1 - STOT RE 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service