All Photos(1)

227137

Sigma-Aldrich

Dibutyl oxalate

99%

Linear Formula:
CH3(CH2)3OCOCOO(CH2)3CH3
CAS Number:
Molecular Weight:
202.25
Beilstein:
1776065
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

99%

refractive index

n20/D 1.423 (lit.)

bp

239-240 °C (lit.)

mp

−29 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)C(=O)OCCCC

InChI

1S/C10H18O4/c1-3-5-7-13-9(11)10(12)14-8-6-4-2/h3-8H2,1-2H3

InChI key

JKRZOJADNVOXPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The rate of dibutyl oxalate degradation in ruminal contents from adapted and non-adapted animals was studied.

Packaging

100 mL in glass bottle

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK Germany

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Changes in ruminal oxalate degradation rates associated with adaptation to oxalate ingestion.
M J Allison et al.
Journal of animal science, 45(5), 1173-1179 (1977-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service