Documents

229482

Sigma-Aldrich

Bismuth(III) oxychloride

≥99.99%

Synonym(s):
Bismuthyl(III) chloride, Chlorobismuth oxide, Bismuth(III) chloride oxide
Empirical Formula (Hill Notation):
BiClO
CAS Number:
Molecular Weight:
260.43
EC Number:
MDL number:
PubChem Substance ID:

assay

≥99.99%

density

7.72 g/mL at 25 °C (lit.)

SMILES string

Cl[Bi]=O

InChI

1S/Bi.ClH.O/h;1H;/q+1;;/p-1

InChI key

GLQBXSIPUULYOG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service