Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
CH3(CH2)3C6H4CO2H
CAS Number:
Molecular Weight:
178.23
EC Number:
243-940-9
UNSPSC Code:
12352103
PubChem Substance ID:
Beilstein/REAXYS Number:
2044046
MDL number:
Skip To
Product Name
4-Butylbenzoic acid, 99%
InChI key
JFKUBRAOUZEZSL-UHFFFAOYSA-N
InChI
1S/C11H14O2/c1-2-3-4-9-5-7-10(8-6-9)11(12)13/h5-8H,2-4H2,1H3,(H,12,13)
SMILES string
CCCCc1ccc(cc1)C(O)=O
assay
99%
form
liquid crystal
mp
100-113 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mette Kristensen et al.
Journal of chromatography. A, 1601, 21-26 (2019-05-13)
Gas-chromatography (GC) analysis of carboxylic acids is limited by the high polarity and low volatility of most of these compounds. Boron trifluoride (BF3) mediated alkylation reactions is one of the most commonly used derivatization methods for making carboxylic acids GC
Dieter Schemeth et al.
Analytica chimica acta, 1038, 182-190 (2018-10-04)
In this study, we focus on isolation and fractionation strategies by solid phase extraction (SPE) for a broad range of environmentally related organic acids. These emerging potential contaminants are primary degradation products of spilled petrogenic compounds but little attention has
Chao-Bin Xue et al.
Bioorganic & medicinal chemistry, 15(5), 2006-2015 (2007-01-30)
Phenoloxidase (PO), also known as tyrosinase, is a key enzyme in insect development, responsible for catalyzing the hydroxylation of tyrosine into o-diphenols and the oxidation of o-diphenols into o-quinones. Inhibition of PO may provide a basis for novel environmentally friendly
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service