MilliporeSigma
All Photos(1)

233773

Sigma-Aldrich

1-(2-Methoxyphenyl)-2-propanol

98%

Linear Formula:
CH3OC6H4CH2CH(OH)CH3
CAS Number:
Molecular Weight:
166.22
MDL number:
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.529 (lit.)

density

1.035 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1CC(C)O

InChI

1S/C10H14O2/c1-8(11)7-9-5-3-4-6-10(9)12-2/h3-6,8,11H,7H2,1-2H3

InChI key

ROVATSABYKBNCG-UHFFFAOYSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service