234419

Sigma-Aldrich

3-Chlorobenzotrifluoride

technical grade, 94%

Synonym(s):
3-Chloro-α,α,α-trifluorotoluene
Linear Formula:
ClC6H4CF3
CAS Number:
Molecular Weight:
180.55
EC Number:
MDL number:
PubChem Substance ID:

grade

technical grade

assay

94%

refractive index

n20/D 1.446 (lit.)

bp

137-138 °C (lit.)

mp

−56 °C (lit.)

density

1.336 g/mL at 25 °C

SMILES string

FC(F)(F)c1cccc(Cl)c1

InChI

1S/C7H4ClF3/c8-6-3-1-2-5(4-6)7(9,10)11/h1-4H

InChI key

YTCGOUNVIAWCMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

remainder mainly 4-isomer

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service