236748

Sigma-Aldrich

2,2′-Bipyridinium chlorochromate

97%

Empirical Formula (Hill Notation):
C10H9ClCrN2O3
CAS Number:
Molecular Weight:
292.64
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

reaction suitability

reagent type: oxidant

SMILES string

O[Cr](Cl)(=O)=O.c1ccc(nc1)-c2ccccn2

InChI

1S/C10H8N2.ClH.Cr.H2O.2O/c1-3-7-11-9(5-1)10-6-2-4-8-12-10;;;;;/h1-8H;1H;;1H2;;/q;;+2;;;/p-2

InChI key

LEZAZQDIYCYEIE-UHFFFAOYSA-L

Signal Word

Danger

hazcat

Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

storage_class_code

5.1B - Oxidizing hazardous materials

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service