237760

Sigma-Aldrich

(S)-(+)-2,4-Diaminobutyric acid dihydrochloride

97%

Synonym(s):
L-2,4-Diaminobutyric acid hydrochloride
Linear Formula:
H2NCH2CH2CH(NH2)CO2H · 2HCl
CAS Number:
Molecular Weight:
191.06
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

optical activity

[α]20/D +12°, c = 1 in H2O

mp

193 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].NCC[C@H](N)C(O)=O

Application

Pharmacological tool and potential chiral building block.

Certificate of Analysis

Certificate of Origin

Tetrahedron Letters, 30, 6011-6011 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service