240206

Sigma-Aldrich

Nicotinamide

≥99%

Synonym(s):
Niacinamide, Vitamin PP, Nicotinic acid amide, Vitamin B3, Pyridine-3-carboxylic acid amide
Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein/REAXYS Number:
383619
EC Number:
MDL number:

assay

≥99%

mp

128-131 °C (lit.)

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Certificate of Analysis

Certificate of Origin

Mitchell E Horwitz et al.
The Journal of clinical investigation, 124(7), 3121-3128 (2014-06-10)
Delayed hematopoietic recovery is a major drawback of umbilical cord blood (UCB) transplantation. Transplantation of ex vivo-expanded UCB shortens time to hematopoietic recovery, but long-term, robust engraftment by the expanded unit has yet to be demonstrated. We tested the hypothesis...
Ann-Lii Cheng et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(32), 4067-4075 (2013-10-02)
Open-label, phase III trial evaluating whether sunitinib was superior or equivalent to sorafenib in hepatocellular cancer. Patients were stratified and randomly assigned to receive sunitinib 37.5 mg once per day or sorafenib 400 mg twice per day. Primary end point...
Salvatore Simmini et al.
Nature communications, 5, 5728-5728 (2014-12-17)
The endodermal lining of the adult gastro-intestinal tract harbours stem cells that are responsible for the day-to-day regeneration of the epithelium. Stem cells residing in the pyloric glands of the stomach and in the small intestinal crypts differ in their...
Kathrin Schmeisser et al.
Nature chemical biology, 9(11), 693-700 (2013-10-01)
Sirtuins, a family of histone deacetylases, have a fiercely debated role in regulating lifespan. In contrast with recent observations, here we find that overexpression of sir-2.1, the ortholog of mammalian SirT1, does extend Caenorhabditis elegans lifespan. Sirtuins mandatorily convert NAD(+)...
Thomas J Herzog et al.
Gynecologic oncology, 130(1), 25-30 (2013-04-18)
Sorafenib, an oral multikinase inhibitor of the VEGFR/PDGFR/Raf/MEK/ERK pathway, has shown potential activity in patients with recurrent ovarian cancer (OC). One strategy to prolong disease control and survival in patients with OC is maintenance therapy after achieving a complete response....

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service