241199

Sigma-Aldrich

2-Bromopropionic acid

≥99%

Synonym(s):
(±)-2-Bromopropionic acid
Linear Formula:
CH3CHBrCOOH
CAS Number:
Molecular Weight:
152.97
Beilstein/REAXYS Number:
1720261
EC Number:
MDL number:
PubChem Substance ID:

assay

≥99%

refractive index

n20/D 1.475 (lit.)

bp

203 °C (lit.)

density

1.7 g/mL at 25 °C (lit.)

SMILES string

CC(Br)C(O)=O

InChI

1S/C3H5BrO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)

InChI key

MONMFXREYOKQTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3261 8 / PGII

WGK Germany

3

Flash Point(F)

212 °F

Flash Point(C)

100 °C

Hari M R Gardimalla et al.
Chemical communications (Cambridge, England), (35)(35), 4432-4434 (2005-09-02)
Candida rugosa lipase immobilized on maghemite nanoparticles demonstrated high stereoselectivity in kinetic resolution of racemic carboxylates and improved long-term stability over its parent free enzyme, allowing the supported enzyme to be repeatedly used for a series of chiral resolution reactions.
J E McGrath et al.
Applied and environmental microbiology, 63(8), 3333-3335 (1997-08-01)
Type strains of the purple nonsulfur species Rhodospirillum rubrum, Rhodospirillum photometricum, and Rhodopseudomonas palustris grew phototrophically on a number of two- and three-carbon halocarboxylic acids in the presence of CO2, by reductive dehalogenation and assimilation of the resulting acid. Strains...
E A Lock et al.
Archives of toxicology, 74(9), 547-554 (2000-12-29)
Oral administration of L-2-chloropropionic acid (L-CPA) to rats either as a single dose (750 mg/kg) or daily doses (250 mg/kg per day for 3 days) produces selective necrosis to the granule cell layer of the cerebellum. As part of a...
O Brandsnes et al.
European journal of biochemistry, 126(2), 247-252 (1982-08-01)
The effect of temperature on the inactivation of liver alcohol dehydrogenase and on the alkylation of a model thiol free in solution by bromoacetate, 2-bromopropionate, 3-bromopropionate and 2-bromo-3-(5-imidazolyl)-propionate has been studied and the thermodynamic activation parameters calculated. All the bromoacids...
Jun Zhao et al.
Dental materials : official publication of the Academy of Dental Materials, 27(5), 478-486 (2011-03-08)
The objective of this study was to synthesize and characterize novel hyperbranched poly(acrylic acid)s via atom-transfer radical polymerization (ATRP) technique and tether the photo-curable methacrylate onto the poly(acrylic acid), use these polymers to formulate the resin-modified glass-ionomer cements, and evaluate...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.