243396

Manganese(II) carbonate

technical grade

Linear Formula:
MnCO3
CAS Number:
Molecular Weight:
114.95
MDL number:

grade

technical grade

density

3.12 g/mL at 25 °C (lit.)

SMILES string

[Mn++].[O-]C([O-])=O

InChI

1S/CH2O3.Mn/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

XMWCXZJXESXBBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service